Learn More
Trajan™ Gas-Tight Autosampler Syringes
Product Code. p-8601032
Click to view available options
Volume (Metric):
5 μL
100 μL
Needle Gauge:
22 G
23 G
Connection Type:
Fixed
Removable
For Use With (Equipment):
CTC, Carlo Erba, Fisons Autosampler
Carlo Erba/Fisons/Leap/CTC ASD200 and AS800
PerkinElmer Autosystem
Unit Size:
Each
Product Code. 12997556
Brand: Trajan™ 005330
Replace syringes for Agilent™ Technologies, Varian™, Perkin Elmer™, Shimadzu™, Carlo Erba™, Fisons™, CTC/Leap™, Hitachi™, and more
Syringe needles are cone point unless otherwise noted.
Order Info
Additional syringes are available for other autosamplers made by the manufacturers represented here, as well as syringes for Unicam™, Waters™, and Siemens™ instruments. For information, contact your Fisher Scientific Customer Service Representative.
Specifications
Needle Gauge 22 G
Diameter (English) Outer Needle 0.02 in.
Diameter (Metric) Outer Needle 0.72 mm
For Use With (Application) High performance liquid chromatology
Length (English) Needle 2 in.
Product Type Autosampler Syringe
Model 100R-C/T-GT-LC
Quantity Ea.
Volume (Metric) 100 μL
Length (Metric) Needle 51 mm
For Use With (Equipment) Carlo Erba/Fisons/Leap/CTC ASD200 and AS800
Connection Type Removable