Learn More
Lenz Laborglasintrumente Gas Inlet Tubes, Straight
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
17.91€ - 24.91€
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | |||||
11836114
|
Lenz Laborglasintrumente
05080002 |
NS 19/26 |
17.91€
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
10284001
|
Lenz Laborglasintrumente
05080003 |
Cone NS 24/29 |
24.91€
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
10070191
|
Lenz Laborglasintrumente
05080004 |
Cone NS 29/32 |
24.17€
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
Description
- Straight air leak tubes
- Built-in length: 200 mm
- With NS cone and thin capillary (easily to draw by burner)
- Made of DURAN™ tubing