Learn More
BOLA Gas Inlet Tubes
Product Code. p-200109975
Click to view available options
Length (Metric):
200 mm
600 mm
Unit Size:
Each
Product Code. 10631332
Brand: BOLA N150202
Only null left
Add to basket
Add to basket
Gas Inlet Tubes M 8 x 1 mm, hose connector 9 mm, Bore 5 mm
  • For constructing a gas washing equipment.
  • Tube with inner diameter 5 mm, one side with hose connector dia. 9 mm for connecting tubing, other side with thread M 8x1 for connecting a gas frit or a gas distributor.
Specifications
Length (Metric) 200 mm