Learn More
BOLA Gas Frits
Product Code. p-200109974
Click to view available options
Connectors:
5 mm
7 mm
6 x 1 mm
Unit Size:
Each
Product Code. 10013481
Brand: BOLA N150328
Gas Frits made of microporous PTFE
  • Microporous PTFE with pore size 5 μm for steady sparkling of the flowing gas.
  • Suitable for scrubber bottles and columns with M 8x1 thread and for gas inlet tubes.
Specifications
Autoclavable Autoclavable
Connectors 6 x 1 mm
Disposable Reusable
Sample Volume 5 μL
Sterility Non-sterile
Length (Metric) 15 mm