Learn More
Asecos™ G-Ultimate 90 Gas Cylinder Cabinet
Product Code. 13423877 Shop All Asecos Products
Change view
Click to view available options
Color:
Light Gray
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
13423877 Light Gray Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13423877 Supplier Asecos™ Supplier No. 3066700230668
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Safe and approved cabinet for storage of flammable or toxic gases in working areas. Asecos™ G-Ultimate 90 Gas Cylinder Cabinet holds up to 3 x 50L gas cylinders.
  • Robust and long-lasting
    • Triple-hinged door
    • Scratch- and impact-resistant
  • Easy handling
    • Doors stop in any position
    • Self-closing in the event of fire
  • Locking doors
  • Easy transport
  • Easy alignment
  • Ventilation
Specifications
Certifications/Compliance FM Approved, 30 EN 14470-1, EKS / AK4 / EN 14727, CE
Configuration 3 Tray Shelves and Bottom Tray, 4 Full Extension Drawers, 6 Full Extension Drawers
Depth (Metric) Exterior 616 mm
Dimensions (D x W x H) 616 x 1196 x 1968 mm
For Use With (Application) Storage of hazardous (flammable) substances in the workplace
Height (Metric) Exterior 1968 mm
Width (Metric) Exterior 1196 mm
Color Light Gray
Product Type Fire safety storage cabinet