Learn More
Wld Tec™ Fuego Pro SCS Gas Burner
Código de producto. 12695670
Cambiar vista
Click to view available options
Product Type:
Laboratory Gas Burner
Tamaño de la unidad:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Product Type Tamaño de la unidad
12695670 Laboratory Gas Burner Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 12695670 Proveedor Wld Tec™ N.º de proveedor 8.204.000
  • ONLINE EXCLUSIVE PRICE
    990.15€
    780.00€ / Pack of 1
    Save 210.15€
    21% Off
 Restricted Item
Estimated Shipment: 20-11-2026
Añadir a la cesta
Añadir a la cesta
Este artículo no se puede devolver. Vea la política de devoluciones
Safe, streamlined, versatile and energy-efficient laboratory gas burner. Wld Tec™ Fuego Pro SCS Gas Burner is fabricated of stainless steel and heat-resistant glass.
  • Button or foot pedal operation
  • Gas inlet filter
  • DVGW approved
  • Animated graphic display
  • Battery operation option
  • Gas consumption display
  • Modern design
    • Low casing promotes working comfort
    • Reduced air distrubance
    • Removable burner head for cleaning
    • Integrated drain
  • Universal
    • Accepts stationary natural gas and propane / butane gas supplies, cartridge gas, or gas cylinders
  • Safe
    • Overheating protection
    • Continuous burner head control
    • Burner head clogging detection
    • Residual heat display
    • Automatic cut-off function prevents unintentional ignition of the flame when the burner has not been ignited for a longer period
    • Acoustic signals
  • Environmentally conscious
    • Energy efficiency in accordance with the most recent standards
  • Take-back guarantee
    • Older WLD-TEC burners will be taken back and disposed of in an environmentally compatible manner
  • Robust
    • Stainless steel construction extends to the controls and gas connection
    • Heat-resistant glass display
Note: Foot pedal is not included; sold separately (see 10263283).
Especificaciones
Gas Type Butane, Natural Gas, Propane
Length (Metric) 103 mm
Material Stainless Steel
Height (Metric) 49 mm
Max. Temperature (Metric) 1300°C (Natural Gas), 1350°C (Liquid Gas)
No. per Pack 1 Ea.
Weight (Metric) 700 g
Width (Metric) 130 mm
Product Type Laboratory Gas Burner
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.