missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Garcinol
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
- Garcinol is an inhibitor of histone acetyltransferases (HATs) p300 and pCAF (IC50 = 7 and 5 μM, respectively) (Balasubramanyam et al.) It also inhibits the HAT GCN5 in Cryptococcus neoformans, inducing temperature sensitivity and impairing growth (O'Meara et al.)
- Promotes ex vivo expansion of human hematopoietic stem cells (Nishino et al.)
- Promotes neurogenesis in rat cortical progenitor cells (Weng et al.)
- Induces apoptosis in several types of cancer cells and has anti-inflammatory actions (Koeberle et al.; Prasad et al.)
Specifications
Specifications
| CAS | 78824-30-3 |
| Synonym | Camboginol |
| Quantity | 5 mg |
| Molecular Formula | C38H50O6 |
| Purity | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction