Learn More
Garcin™ Sanicid 5™ Detergent
Product Code. 11708733
Click to view available options
pH:
9.3
9.6
Unit Size:
5 L
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11708733
Brand: Garcin™ LB922700
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Use Garcin™ Sanicid 5™ Detergent is ideal for cleaning elementary spills and soil that occur in a laboratory.
  • Liquid
  • Safe from Light at > 4 ° C
  • Anionic and chlorinated Pure pH: 10 ± 0.5 • pH diluted to 2%: 9.3 ± 0.5
  • Expiration: 4 years
Specifications
Product Type Sanicid 5 Detergent
Form Liquid
pH 9.3
Appearance Colorless
Certifications/Compliance NF EN 1040, EN 1276, NF T 72-151 and NF T spectrum 5 72-171, NF EN 1275, NF T 72-180
Container Type Can
Content And Storage >4°C
Volume (Metric) 5 L
compliance-icons compliance-icons
Product Identifier
  • SANICID 5 INODORE 5L
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P391-Collect spillage.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : didecyldimethylammonium chloride, benzalkonium chlroide, propan-2-ol, isotridecanol ethoxylated