Learn More
Garcin™ Sanicid 5™ Detergent
Product Code. 11703574
Change view
Click to view available options
pH:
9.3
9.6
Unit Size:
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH unitSize
11703574 9.6 5 L
11708733 9.3 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11703574 Supplier Garcin™ Supplier No. LB922800
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Use Garcin™ Sanicid 5™ Detergent is ideal for cleaning elementary spills and soil that occur in a laboratory.
  • Liquid
  • Safe from Light at > 4 ° C
  • Anionic and chlorinated Pure pH: 10 ± 0.5 • pH diluted to 2%: 9.3 ± 0.5
  • Expiration: 4 years
Specifications
Product Type Sanicid 5 Perfumed Detergent
Form Liquid
pH 9.6
Appearance Amber
Certifications/Compliance NF EN 1040, EN 1276, NF T 72-151 and NF T spectrum 5 72-171, NF EN 1275, NF T 72-180
Container Type Can
Content And Storage 4°C
Volume (Metric) 5 L
compliance-icons compliance-icons
Product Identifier
  • SANICID 5 PARFUME 5 L
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: didecyldimethylammonium chloride, benzalkonium chloride, propan-2-ol, isotridecanol ethoxylated