Learn More
Garcin™ Bactinyl Disinfectant Detergent
Product Code. 11788743
Change view
Click to view available options
Volume (Metric):
500 mL
5 L
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
11788743 500 mL Each
11798723 5 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11788743 Supplier Garcin™ Supplier No. LB920723
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Use Garcin™ Bactinyl Disinfectant Detergent as a janitorial cleaner and disinfectant. Its active substances include peroxides, quaternary ammoniums and ethanol.
  • Sterile
  • Rinse not justified if thorough drying
  • Odorless
  • Inactivation of the HIV virus in 5 minutes
  • Approved for food contact
Specifications
Product Type Bactinyl Disinfectant Detergent
Form Liquid
pH 10 ±1
Appearance Opalescent
Certifications/Compliance EN 1040, EN 1276, NF T 72-151 spectrum 5, NF T 72-190 NF T 72-171et, NF EN 1275 and NF EN 1650, NF T 72-180, EC 0398
Container Type Bottle
Content And Storage 4°C
Quantity 1 Ea.
Volume (Metric) 500 mL
compliance-icons
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P273-Avoid release to the environment.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.