Learn More
Garcin™ Algaecide Bath
Product Code. 11778743
Change view
Click to view available options
Volume (Metric):
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
11778743 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11778743 Supplier Garcin™ Supplier No. LB920410
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Garcin™ Algaecide Bath prevents the proliferation of bacteria, algae and fungi.
  • Bactinyl™ algaecide
  • Prevents the proliferation of bacteria, algae and fungi
  • Without chlorinated products or solvents
  • Use without risk of altering the regulation systems
  • Solubilizing and descaling properties
  • Contains an anti-foaming agent
  • Dilution at 1%, renewal every 8 days
  • Prevents the development of microscopic algae
  • Not recommended for water tank CO2 incubators
  • Algaecide Bactinyl recommended
Specifications
Dilution Dilution at 1%, renewal every 8 days
Includes Contains an anti-foaming agent
Quantity 1 Ea.
Volume (Metric) 250 mL
compliance-icons
Product Identifier
  • bactinyl bain marie
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P273-Avoid release to the environment.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: dimethylbenzylammonium chloride