Learn More
Thermo Scientific™ gamma-Secretase Inhibitor II
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Quantity | 5 mg |
| Description | Off-white |
| Content And Storage | -30°C to -10°C |
| For Use With (Application) | A cell-permeable, reversible and selective peptidomimetic inhibitor of --secretase |
| Molecular Formula | C20H30N2O4 |
| Formula Weight | 362.47 |
| Synonym | MW167 |
| Product Type | gamma-Secretase Inhibitor |
| Form | Solid |