Learn More
Fisherbrand™ Gamma Radiation Safe Midi Storage Box
Product Code. 12327107 Shop All Fisherbrand Products
Change view
Click to view available options
Dimensions (L x W x H):
16.5 x 8.5 x 6 cm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) unitSize
12327107 16.5 x 8.5 x 6 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12327107
Estimated Shipment: 01-10-2026
Add to basket
Add to basket
Provides radiation protection shielding for use in laboratories.
  • Manufactured with hinged lids
  • Transparent shielding
  • Serves as solution for short-term storage of radioactive waste or radioisotopes
  • Constructed of acrylic provides easy fabrication and manufacturability
Specifications
Dimensions (L x W x H) 16.5 x 8.5 x 6 cm
For Use With (Application) Gamma radiation shielded
Holds Radioisotopes or radioactive waste
Includes Hinged lid
Product Type Midi Storage Box
Material Acrylic