Learn More
Fisherbrand™ Gamma Radiation Safe Block for Eppendorf Tubes
Product Code. 12357137 Shop All Fisherbrand Products
Change view
Click to view available options
Dimensions (L x W x H):
3.5 x 14 x 5 cm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) unitSize
12357137 3.5 x 14 x 5 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12357137
Estimated Shipment: 13-11-2026
Add to basket
Add to basket
Provides radiation protection shielding for use in laboratories.
  • Transparent shielding
  • Serves as solution for short-term storage of radioactive waste or radioisotopes
  • Constructed of acrylic provides easy fabrication and manufacturability
Specifications
Dimensions (L x W x H) 3.5 x 14 x 5 cm
For Use With (Application) Gamma Radiation Safe
Holds 4 x 1.5mL Eppendorf tubes
Product Type Block for Eppendorf tubes
Material Acrylic