Learn More
Gamma-Butyrobetaine-d9 Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | gamma-Butyrobetaine trimethyl-d9 hydrochloride |
| CAS | 85806-17-3 |
| Molecular Formula | C7 2H9 H7 N O2 . Cl |
| Quantity | 5 mg |
| Synonym | 1-Propanaminium, 3-carboxy-N,N,N-tri(methyl-d3)-, chloride (1:1) (ACI), 1-Propanaminium, 3-carboxy-N,N,N-tri(methyl-d3)-, chloride (9CI), 3-Carboxy-N,N,N-tri(methyl-d3)-1-propanaminium chloride, N-(Carboxypropyl)-N,N,N-trimethyl-d9-ammonium Chloride, γ-Butyrobetaine-d9 Hydrochloride, (3-Carboxypropyl)trimethylammonium-d9 Chloride, (3-Carboxypropyl)trimethylammonium-D9 (N,N,N-trimethyl-D9) Chloride |
| InChI Formula | InChI=1S/C7H15NO2.ClH/c1-8(2,3)6-4-5-7(9)10;/h4-6H2,1-3H3;1H/i1D3,2D3,3D3; |
| SMILES | [Cl-].[2H]C([2H])([2H])[N+](CCCC(=O)O)(C([2H])([2H])[2H])C([2H])([2H])[2H] |
| IUPAC Name | 3-carboxypropyl-tris(trideuteriomethyl)azanium;chloride |
| Molecular Weight (g/mol) | 190.72 |
| Formula Weight | 190.1434 |