Learn More
Rogo Sampaic™ Galvanized Steel Opened Rings
Click to view available options
Diameter (Metric) Outer:
60 mm
80 mm
100 mm
140 mm
Unit Size:
Each
Description
- Available in a range of diameters
- Helps support a variety of glassware and other chemistry equipment
Specifications
Specifications
| Material | Steel |
| Diameter (Metric) Outer | 60 mm |
| Product Type | Opened Ring |
| Quantity | 1 Ea. |