Learn More
asecos™ Galvanised Steel Spray Can
Shop All Asecos Products
Click to view available options
Capacity (Metric):
1 L
2 L
Color:
Gray
Yellow
Unit Size:
Each
Description
- Spray attachment made of plastic with adjustable nozzle.
- Integrated plastic pump.
- Incl. multilingual safety sticker to designate the contents.
Specifications
Specifications
| Product Type | Spray Can |
| Capacity (English) | 0.5 gal. |
| Capacity (Metric) | 2 L |
| Color | Yellow |
| For Use With (Application) | Spray cleaning surfaces sparingly and in measured doses |
| Material | Steel |
| Width (English) | 7.40 in. |
| Width (Metric) | 188.00 mm |
| Height (English) | 13.58 in. |
| Height (Metric) | 345.00 mm |
| Show More |