Learn More
asecos™ Galvanised Steel Spray Can
Product Code. 17856446 Shop All Asecos Products
Change view
Click to view available options
Capacity (Metric):
1 L
2 L
Color:
Gray
Yellow
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Color unitSize
17856446 1 L Yellow Each
17876566 2 L Yellow Each
17846356 1 L Gray Each
17886536 2 L Gray Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17856446 Supplier asecos™ Supplier No. 23137
This item requires a price update prior to being ordered.
Request a Price for this item here.
The Galvanised Steel Spray Can is ideal for economical and controlled spraying of surfaces and parts to be cleaned.
  • Spray attachment made of plastic with adjustable nozzle.
  • Integrated plastic pump.
  • Incl. multilingual safety sticker to designate the contents.
Specifications
Product Type Spray Can
Capacity (English) 0.3 gal.
Capacity (Metric) 1 L
Color Yellow
For Use With (Application) Spray cleaning surfaces sparingly and in measured doses
Material Steel
Width (English) 6.38 in.
Width (Metric) 162.00 mm
Height (English) 11.81 in.
Height (Metric) 300.00 mm
Weight (English) 1.65 lb.
Weight (Metric) 0.75 kg