missing translation for 'onlineSavingsMsg'
Learn More

Gallein, TRC

Product Code. 30392945
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392945 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30392945 Supplier Toronto Research Chemicals Supplier No. TRCG190553100MG
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Gallein
CAS 2103-64-2
Molecular Formula C20H12O7
Quantity 100 mg
Synonym 3',4',5',6'-Tetrahydroxyspiro[isobenzofuran-1(3H),9'-(9H)xanthen]-3-one, Alizarine Violet, C.I. 45445, C.I. Mordant Violet 25, Mordant Violet 25, NSC 56445, NSC 622478, NSC 8668, Pyrogallolphthalein
InChI Formula InChI=1S/C20H12O7/c21-13-7-5-11-17(15(13)23)26-18-12(6-8-14(22)16(18)24)20(11)10-4-2-1-3-9(10)19(25)27-20/h1-8,21-24H
SMILES Oc1ccc2c(Oc3c(O)c(O)ccc3C24OC(=O)c5ccccc45)c1O
IUPAC Name 3',4',5',6'-tetrahydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one
Molecular Weight (g/mol) 364.31
Formula Weight 364.0583
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.