Learn More
Eppendorf™ Galaxy™ 48 R Incubators
Product Code. 15594439
Click to view available options
Door Style:
2-Split Inner Door
Single Glass Inner Door
Oxygen Control:
1 to 19% O2 Control
None
Unit Size:
Each
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Product Code. 15594439
Brand: Eppendorf™ CO48312041
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
When a high level of cell culture isolation is required or very limited space is available in the lab, the stackable Galaxy 48R is the right choice. The high temperature disinfection, a seamless chamber and a fanless design makes it easy to clean and provides high contamination protection.
  • 48 Liter (1.7 cu. ft.) provides mid-sized capacity within a small footprint
  • Options: High Temp. Disinfection (4 hour at 120 °C), OO2control (e.g. for hypoxic studies), 2-split inner glass door
  • Fanless design offers more usable space
  • Six-sided direct heating system provides a uniform incubated environment
  • Deep-drawn, easy-to-clean stainless steel chamber without seams or welds
  • Low gas consumption
  • Integrated alarm relay
TRUSTED_SUSTAINABILITY
Specifications
Capacity (English) 1.7 cu. Ft.
CO2 Concentration Control ± 0.1%
Description UK/HKG Power Supply, High Temperature Disinfection
Capacity (Metric) 48 L
Chamber Material Stainless Steel
CO2 Concentration Range ±0.2 to 20%
CO2 Sensor Technology IR Sensor
No. of Shelves 6
O2 Concentration Range 0.1 to 19%
Oxygen Control 1 to 19% O2 Control
Temperature Range (Metric) Ambient +4° to 50°C
Type CO2 Incubator
Disinfection Time 4 hours at 120°C
Door Style 2-Split Inner Door
Ports RS-232 Communication Port, 25mm Access Ports
Dimensions (L x W x H) 47.5 x 48.3 x 64.5 cm
Length (Metric) 47.5 cm
Width (Metric) 48.3 cm
Height (Metric) Interior 64.5 cm
Weight (Metric) 31.8 kg
Electrical Requirements 230 V, 50/60 Hz
Voltage 230 V
Frequency 50/60 Hz