missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Galanin (1-30) (human)

Product Code. 18054812
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 18054812

Brand: Tocris Bioscience™ 1179/1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Endogenous galanin receptor agonist

Galanin (1-30) (human) is a endogenous peptide with multiple endocrine, metabolic and behavioral effects. Has been shown to have an action on intestinal smooth muscle, insulin and somatostatin release, and synaptic neurotransmission.
TRUSTED_SUSTAINABILITY

Specifications

Species Human
Purity 95%
CAS 119418-04-1
Content And Storage Store at -20°C
Molecular Weight (g/mol) M.W. (Observed): 3157.41
Quantity 1 mg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.