missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gadobutrol, TRC
Specifications
Specifications
| Chemical Name or Material | Gadobutrol |
| CAS | 770691-21-9 |
| Molecular Formula | C18 H31 Gd N4 O9 |
| Quantity | 5 g |
| Synonym | Gadolinium, [rel-10-[(2R,3S)-2-(hydroxy-κO)-3-hydroxy-1-(hydroxymethyl)propyl]-1,4,7,10-tetraazacyclododecane-1,4,7-triacetato(3-)-κN1,κN4,κN7,κN10,κO1,κO4,κO7]-, (SA-8-1425362'5')-, Gadolinium, [10-[2-(hydroxy-κO)-3-hydroxy-1-(hydroxymethyl)propyl]-1,4,7,10-tetraazacyclododecane-1,4,7-triacetato(3-)-κN1,κN4,κN7,κN10,κO1,κO4,κO7]-, [SA-8-1425362'5'-(R*,S*)]-, Gadobutrol, Gadograf, Gadovist, Protovist, ZK 135079 |
| InChI Formula | InChI=1S/C18H34N4O9.Gd/c23-12-14(15(25)13-24)22-7-5-20(10-17(28)29)3-1-19(9-16(26)27)2-4-21(6-8-22)11-18(30)31;/h14-15,23-25H,1-13H2,(H,26,27)(H,28,29)(H,30,31);/q;+3/p-3/t14-,15-;/m1./s1 |
| SMILES | OC[C@@H](O)[C@@H](CO)N12CCN3(CC(=O)[O-])CCN4(CC(=O)[O-])CCN(CC(=O)[O-])(CC1)[Gd+3]234 |
| Molecular Weight (g/mol) | 604.71 |
| Formula Weight | 605.1332 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?