Learn More
Thermo Scientific™ Gable Cover for VersaCool™ Refrigerated Circulating Bath
Shop All Thermo Scientific ProductsDescription
Utilize the gable cover to accommodate larger size beakers for optimal performance with the Thermo Scientific™ VersaCool™ Refrigerated Circulating Bath.
Optional accessory for the Thermo Scientific™ VersaCool™ Refrigerated Circulating Bath. This reversible hinged cover allows for larger size beakers and:
- Reduces moisture and directs condensate back into bath
- Ideal for water, water/glycol applications
Specifications
Specifications
| For Use With (Equipment) | Thermo Scientific VersaCool Refrigerated Circulating Bath |
| Type | Gable Cover |