missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gabapentin-d4 (aminomethyl-d2; acetic-d2 acid), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Gabapentin-d4 |
| CAS | 1185039-20-6 |
| Molecular Formula | C9 D4 H13 N O2 |
| Quantity | 0.01 g |
| Synonym | Gabapentin-d4, Gabapentin D4 (aminomethyl D2, acetic acid D2), Cyclohexaneacetic-α,α-d2 acid, 1-(aminomethyl-d2)- (ACI), 1-(Aminomethyl-d2)cyclohexaneacetic-α,α-d2 acid (ACI), 2-[1-(Amino-dideuteriomethyl)cyclohexyl]-2,2-dideuterioacetic acid |
| InChI Formula | InChI=1 S/C9H17NO2/c10-7-9(6-8(11)12)4-2-1-3-5-9/h1-7,10H2,(H,11,12)/i6D2,7D2 |
| SMILES | [2 H]C([2 H])(N)C1(CCCCC1)C([2 H])([2 H])C(=O)O |
| IUPAC Name | 2-[1-[amino(dideuterio)methyl]cyclohexyl]-2,2-dideuterioacetic acid |
| Molecular Weight (g/mol) | 175.261 |
| Formula Weight | 175.151 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction