Learn More
STEMCELL Technologies G418
Product Code. 17120002
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17120002 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17120002 Supplier STEMCELL Technologies Supplier No. 03812
 Restricted Item
Estimated Shipment: 25-11-2026
Antibiotic for selecting transfected mammalian cells
  • Antibiotic G418 is an aminoglycoside related to gentamicin and is toxic to bacteria, yeast, higher plants, mammalian cells, protozoans, and helminths
  • G418 binds to 80S ribosomes in prokaryotic and eukaryotic cells and inhibits protein synthesis, resulting in cell death
  • Resistance to the antibiotic is conferred by aminoglycoside-3-phosphotransferase
  • Vectors containing the G418 resistance gene (neo) are commonly used for transfecting eukaryotic cells, with selection of transfected cells achieved by culturing them in the presence of G418
  • Cells may exhibit variable resistance to the antibiotic and can take up to a week to die
  • The G418 antibiotic is suitable for use in cell culture for the selection and maintenance of G418-resistant clones when using ClonaCell™-TCS Medium (Catalog #03814), ClonaCell™-CHO CD Medium (Catalog #03815), ClonaCell™-CHO ACF Medium (Catalog #03816), ClonaCell™­CHO CD Liquid Medium (Catalog #03817), or other cell culture media
Specifications
CAS 108321-42-2