Learn More
HUBER Manually Adjustable Bypass G3/4
Click to view available options
For Use With (Equipment):
Unichiller
Unit Size:
Each
Specifications
Specifications
| For Use With (Equipment) | Unichiller |
| Temperature Range | -20°C to +150°C |
| Type | Manually Adjustable Bypass |