Learn More
Novus Biologicals™ g-Linked ATP Agarose Affinity Resin
Affinity resin for purification of enzymes that bind nucleotides and related molecules
Brand: Novus Biologicals™ 510-0002
Description
Resins in which ATP is linked via the gamma-phosphate have been valuable in identifying proteins in the purine-binding proteome, which includes kinases, heat shock proteins and other ATP-binding proteins.
- ATP Agarose resin comprises ATP attached to agarose beads via its gamma-phosphate
- Two forms of resin are available with low and high ligand substitution
- Long hydrophilic spacer (14-atom) minimises unwanted hydrophobic interactions and facilitates unhindered interactions with biomolecules
Recommended Storage
Store at 4°C. Do not freeze.
Specifications
| High ligand substitution (8-12μmol/mL) | |
| 4°C. Do not freeze. | |
| 2 mL | |
| g-Linked ATP Agarose Affinity Resin |