Learn More
Novus Biologicals™ g-Linked ATP Agarose Affinity Resin
Affinity resin for purification of enzymes that bind nucleotides and related molecules
Brand:  Novus Biologicals™ 510-0002
Encompass
Product Code. 15734107
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
Resins in which ATP is linked via the gamma-phosphate have been valuable in identifying proteins in the purine-binding proteome, which includes kinases, heat shock proteins and other ATP-binding proteins.
  • ATP Agarose resin comprises ATP attached to agarose beads via its gamma-phosphate
  • Two forms of resin are available with low and high ligand substitution
  • Long hydrophilic spacer (14-atom) minimises unwanted hydrophobic interactions and facilitates unhindered interactions with biomolecules
Recommended Storage
Store at 4°C. Do not freeze.
Specifications
Specifications
High ligand substitution (8-12μmol/mL)
4°C. Do not freeze.
2 mL
g-Linked ATP Agarose Affinity Resin