Learn More
A&D Weighing FX/FZ Precision Balances IP 65
A&D Weighing FZ/FX Series Precision Balances combine all of the essential weighing functions in a compact, light-weight package.
1119.20€ - 1349.39€
Specifications
Certifications/Compliance | GLP/GMP/GCP/ISO/IP65 |
---|---|
Connection Type | RS-232 C, Quick USB (option), LAN (option) |
Material | Stainless Steel |
Stabilization Time | 1 second |
Electrical Requirements | 11 VA to the AC adaptor (12 V, 1 A) |
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
16631072
|
A&D Weighing
FX-2000I-WP |
2200 g |
1284.57€
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
|||||
16671072
|
A&D Weighing
FX-3000I-WP |
3200 g |
1349.39€
Each |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
16691062
|
A&D Weighing
FX-1200I-WP |
1220 g |
1219.75€
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
|||||
16691072
|
A&D Weighing
FX-300I-WP |
320 g |
1349.39€
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
|||||
16651072
|
A&D Weighing
FX-200I-WP |
220 g |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Description
- IP65 water / dust proof
- Built-in internal calibration at the touch of a button
- Shock absorber function
- Auto power ON/OFF function
- B5 size footprint for use in narrow spaces
- VFD Display for easy to read weighing results
- SCF – Statistical Calculation Function
- Standard RS-232C
- Fast compact SHS sensor, response time only 1 second
- Quick USB interface – no driver installation required (optional)
- LAN – Ethernet interface with WinCT-Plus software (optional)
- Animal weighing pan (optional)
- Large breeze break (optional)
- Small breeze break included on 1 mg models
- Built-in rechargeable NiMH battery pack (optional)
Specifications
GLP/GMP/GCP/ISO/IP65 | |
Stainless Steel | |
11 VA to the AC adaptor (12 V, 1 A) |
RS-232 C, Quick USB (option), LAN (option) | |
1 second |