Learn More
A&D Weighing FX/FZ Precision Balances IP 65
Click to view available options
Capacity:
220 g
320 g
1220 g
2200 g
3200 g
Unit Size:
Each
Description
- IP65 water / dust proof
- Built-in internal calibration at the touch of a button
- Shock absorber function
- Auto power ON/OFF function
- B5 size footprint for use in narrow spaces
- VFD Display for easy to read weighing results
- SCF – Statistical Calculation Function
- Standard RS-232C
- Fast compact SHS sensor, response time only 1 second
- Quick USB interface – no driver installation required (optional)
- LAN – Ethernet interface with WinCT-Plus software (optional)
- Animal weighing pan (optional)
- Large breeze break (optional)
- Small breeze break included on 1 mg models
- Built-in rechargeable NiMH battery pack (optional)
Specifications
Specifications
| Certifications/Compliance | GLP/GMP/GCP/ISO/IP65 |
| Connection Type | RS-232 C, Quick USB (option), LAN (option) |
| Capacity | 220 g |
| Display Type | VFD |
| Readability | 0.001 g |
| Material | Stainless Steel |
| Model | FX-200i-WP |
| Stabilization Time | 1 second |
| Electrical Requirements | 11 VA to the AC adaptor (12 V, 1 A) |