Learn More
Asecos™ FX-CLASSIC-90 Recirculating Air Safety Storage Cabinet
Product Code. 15630159 Shop All Asecos Products
Change view
Click to view available options
Color:
Anthracite Gray, Light Gray
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
15630159 Anthracite Gray, Light Gray Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15630159 Supplier Asecos™ Supplier No. 3328604133656
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
For the safe and approved storage of flammable liquids and further hazardous substances with air circulation for the extraction and filtering of vapors that are harmful to health.
  • Safety for employees due to avoidance of explosive atmospheres
  • Fire resistance of 90 minutes
  • Maximum flexibility in installation, storage and available interior equipment
  • Recirculating air filter cabinets enable the common storage, extraction and filtration of the most diverse hazardous materials in one cabinet
  • Electronic monitoring for filter saturation, air recirculation rate, service intervals and manual filter check
  • Eco-friendly with long filter lifetimes and reusable filter system
  • Possible interior equipment includes drawers, height-adjustable shelves and bottom collecting sumps
  • Robust construction and longevity – triple-hinged doors, safety elements assembled outside of storage compartments for increased corrosion protection, scratch- and impact-resistant surface and easy to clean
  • Doors stop in any position and are self-closing in the event of fire
  • No unauthorized use – doors lockable with cylinder locking (integration into an existing locking system possible) with locking state indicator (red/green)
  • In accordance with EN 14470-1
  • Easy transport with integrated base with removable, optional base cover
  • Easy alignment with adjusting aids to compensate for uneven floors
  • Ideal for the direct installation in working areas with a very low noise level, approx. 39 dB
  • With multi-stage wideband filter
Specifications
Certifications/Compliance EN 14470-1
Configuration 3 x powder-coated sheet steel shelves, load capacity 75 kg; 1 x powder-coated sheet steel perforated insert, load capacity 75 kg; 1 x powder-coated sheet steel bottom collecting sump, capacity 22 L; 3 x PP tray shelves, load capacity 50 kg, capacity 11 L; 1 x PP bottom collecting sump, capacity 11 L
Dimensions (D x W x H) Interior 522 x 450 x 1647 mm
Dimensions (D x W x H) 615 x 1193 x 2294 mm
Door Style Fire-rated (1.5 hr.)
For Use With (Application) Storage of flammable liquids and hazardous substances
Includes Air circulation, filtering, extraction
Load Bearing Capacity 600 kg
No. of Doors 2
Weight (Metric) 565.5 kg
Color Anthracite Gray, Light Gray
Material Gypsum
Product Type Flammables Storage Cabinet