missing translation for 'onlineSavingsMsg'
Learn More

Fursultiamine Hydrochloride, TRC

Product Code. 30012568
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012568 250 mg 250mg
30004201 100 mg 100mg
30004829 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012568 Supplier Toronto Research Chemicals Supplier No. TRCF865230250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2105-43-3
Molecular Formula C17H27ClN4O3S2
Molecular Weight (g/mol) 435
Synonym N-[(4-Amino-2-methyl-5-pyrimidinyl)methyl]-N-[4-hydroxy-1-methyl-2-[(tetrahydrofurfuryl)dithio]-1-butenyl]-formamide Monohydrochloride, N-[(4-Amino-2-methyl-5-pyrimidinyl)methyl]-N-[4-hydroxy-1-methyl-2-[[(tetrahydro-2-furanyl)methyl]dithio]-1-butenyl]-formamide Monohydrochloride, Fursultiamine Hydrochloride, TTFD Hydrochloride, Thiamin Tetrahydrofurfuryl Disulfide Hydrochloride, Thiamine Tetrahydrofurfuryl Disulfide Hydrochloride
IUPAC Name N-[(4-amino-2-methylpyrimidin-5-yl)methyl]-N-[(E)-5-hydroxy-3-(oxolan-2-ylmethyldisulfanyl)pent-2-en-2-yl]formamide;hydrochloride
SMILES CC1=NC=C(C(=N1)N)CN(C=O)C(=C(CCO)SSCC2CCCO2)C.C

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C17H26N4O3S2.ClH/c1-12(16(5-6-22)26-25-10-15-4-3-7-24-15)21(11-23)9-14-8-19-13(2)20-17(14)18;/h8,11,15,22H,3-7,9-10H2,1-2H3,(H2,18,19,20);1H/b16-12+;
Formula Weight 434.121
Percent Purity >95%
Chemical Name or Material Fursultiamine hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.