Learn More
Carbolite™ Furnace with Uniform Temperature Envelope
Product Code. 10704923
Click to view available options
Capacity (Metric):
15 L
23 L
Temperature (Metric):
1200°C
1300°C
1600°C
Dimensions (D x W x H) Interior:
305 x 220 x 220 mm
400 x 245 x 235 mm
Unit Size:
Each
Product Code. 10704923
Brand: Carbolite™ CWF1223230SN+&02301
Only null left
Specifications
Controller Type 301 PID Controller
Capacity (Metric) 23 L
Temperature (Metric) 1200°C
Type Furnace
Dimensions (D x W x H) Interior 400 x 245 x 235 mm
Width (Metric) 505 mm
Width (Metric) Interior 254 mm
Height (Metric) 705 mm
Height (Metric) Interior 235 mm
Depth (Metric) 675 mm
Depth (Metric) Interior 400 mm
Electrical Requirements 230 V 50/60 Hz 7000 w
Voltage 230 V
Wattage 7000 w
Frequency 50/60 Hz