Learn More
Furaneol (technical grade), TRC
Specifications
Specifications
| Chemical Name or Material | 2,5-Dimethyl-4-hydroxy-3(2H)-furanone (Technical Grade) |
| CAS | 3658-77-3 |
| Molecular Formula | C6H8O3 |
| Quantity | 10 g |
| Synonym | 2,5-Dimethyl-4-hydroxy-3(2H)-furanone (Technical Grade), 4-Hydroxy-2,5-dimethyl-3(2H)-furanone, 2,5-Dimethyl-3-hydroxy-4-oxo-4,5-dihydrofuran, 2,5-Dimethyl-4,5-dihydrofuran-3-ol-4-one, 2,5-Dimethyl-4-hydroxy-2,3-dihydrofuran-3-one, 4-Hydroxy-2,5-dimethyl-3-oxo-2,3-dihydrofuran, 4-Hydroxy-2,5-dimethyl-2,3-dihydrofuran-3-one, 4-Hydroxy-2,5-dimethyl-2H-furan-3-one, 4-Hydroxy-2,5-dimethyl-3(2H)-furanone, Alletone, Furaneol, Pineapple Ketone |
| InChI Formula | InChI=1S/C6H8O3/c1-3-5(7)6(8)4(2)9-3/h3,8H,1-2H3 |
| SMILES | CC1OC(=C(O)C1=O)C |
| IUPAC Name | 4-hydroxy-2,5-dimethylfuran-3-one |
| Molecular Weight (g/mol) | 128.13 |
| Formula Weight | 128.0473 |