missing translation for 'onlineSavingsMsg'
Learn More

Furaltadone Hydrochloride, TRC

Product Code. 30393398
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393398 25 g 25g
30402598 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30393398 Supplier Toronto Research Chemicals Supplier No. TRCF86340025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Furaltadone hydrochloride
CAS 3759-92-0
Molecular Formula C13 H16 N4 O6 . Cl H
Quantity 25 g
Synonym 2-Oxazolidinone, 5-(4-morpholinylmethyl)-3-[[(5-nitro-2-furanyl)methylene]amino]-, hydrochloride (1:1), 2-Oxazolidinone, 5-(4-morpholinylmethyl)-3-[[(5-nitro-2-furanyl)methylene]amino]-, monohydrochloride (9CI), 2-Oxazolidinone, 5-(morpholinomethyl)-3-[(5-nitrofurfurylidene)amino]-, monohydrochloride (8CI), 2-Oxazolidinone, 5-morpholinomethyl-3-(5-nitrofurfurylideneamino)-, hydrochloride (6CI), Furaltadone hydrochloride, NF 269, NF-902 hydrochloride, NSC 53270
InChI Formula InChI=1S/C13H16N4O6.ClH/c18-13-16(14-7-10-1-2-12(22-10)17(19)20)9-11(23-13)8-15-3-5-21-6-4-15;/h1-2,7,11H,3-6,8-9H2;1H
SMILES Cl.[O-][N+](=O)c1oc(C=NN2CC(CN3CCOCC3)OC2=O)cc1
IUPAC Name 5-(morpholin-4-ylmethyl)-3-[(5-nitrofuran-2-yl)methylideneamino]-1,3-oxazolidin-2-one;hydrochloride
Molecular Weight (g/mol) 360.75
Formula Weight 360.0837
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.