Description
- Safety funnels based on the SCAT™ 'ARNOLD' funnel series
- Ready-to-use sets consisting of safety funnels and barrel adapters
- Made of high-quality plastics for durable and long-lasting use
- Adapters can be combined with all GL45 safety funnels of the 'ARNOLD' and 'MARCO' series
- Available for all common thread types and sizes
- Also available in electrically conductive material for hazardous areas
Specifications
Specifications
| Capacity (Metric) | 650 mL |
| Closure Type | Hinged Lid |
| Color | Black |
| Height (Metric) | 210 mm |
| Includes | Lid, Splash Guard, Removable Sieve, Grounding Cable |
| For Use With (Equipment) | SCAT ARNOLD Funnel |
| Material | HDPE-EC |
| Product Type | Safety Funnel |