Learn More
Bochem™ Funnels
Product Code. p-8006742
Click to view available options
Diameter (Metric) Stem:
17/13mm
20mm
Height (Metric):
115 mm
135 mm
Unit Size:
Each
Product Code. 10383811
Brand: Bochem™ 8840
Made from 18/10 stainless steel. The Bochem Funnels are used to work with powders.
  • Special funnel
  • For powder
  • Straight tube
  • Made from 18/10 stainless steel
Specifications
Diameter (Metric) Stem 17/13mm
Material Steel
Height (Metric) 115 mm