Description
- Spare reducing inserts for Buerkle™ Funnel stands
- Red
- Round
- PVC
Specifications
Specifications
| Material | PVC |
| Product Type | Insert |
| Shape | Round |
| Color | Red |
| For Use With (Equipment) | Funnel Stands |
| Quantity | 2/Pk. |
| Weight (Metric) | 0.0643 kg |