Learn More
SCAT™ Funnel with Lid and Stainless Steel Strainer
Product Code. 10725602 Shop All SCAT Europe Products
Change view
Click to view available options
Includes:
Lid and Strainer
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
10725602 Lid and Strainer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10725602 Supplier SCAT™ Supplier No. 108670
  • ONLINE EXCLUSIVE PRICE
    243.00€
    222.28€ / Pack of 1
    Save 20.72€
    8% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Open containers create fire and health hazards. Collect liquid waste safely and according to the latest state of the art: This funnel is characterised by its large diameter of 250 mm.
  • The stainless steel strainer safely catches solids such as magnetic stirring bars.
  • With male thread R2 BSP/G2 / BCS 56x4
Specifications
Diameter (Metric)Top 250 mm
Capacity (Metric) 330 mL
Material HDPE
Height (Metric) 240 mm
Color Black
Description Funnel made of PE-HD in red with sieve from stainless steel
Includes Lid and Strainer
Product Type Safety Funnel