Learn More
Afora™ Funnel for General Use
Product Code. 12058892
Change view
Click to view available options
Capacity (Metric):
3 mL
6 mL
10 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
12058892 6 mL Each
12018892 10 mL Each
12038892 3 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12058892 Supplier Afora™ Supplier No. 6395/6
  • ONLINE EXCLUSIVE PRICE
    57.03€
    48.54€ / Pack of 1
    Save 8.49€
    14% Off
Estimated Shipment: 20-11-2026
Prevent spillage with the Afora™ Funnel for General Use. A funnel is a pipe with a wide (often conical) mouth and a narrow stem.
  • Glass
Specifications
Capacity (Metric) 6 mL
Material Glass
Product Type Funnel