Learn More
SCAT™ Safety Funnel 'ARNOLD' with Ball and Lid, V2.0, HDPE-EC
Product Code. 17298860 Shop All SCAT Europe Products
Change view
Click to view available options
Thread Size (Metric):
B83
GL45
R2
S50
S51
S55
S60/61
S65
S70/71
S90
S95
Includes:
Splash Guard, 2 x Removable Sieves, Grounding Cable
Splash Guard, Sieve, Grounding Cable
Splash Guard, Sieve, Grounding Cable, Ball Valve
Splash Guard, Sieve, Grounding Cable, Ball Valve, Lance
For Use With (Equipment):
Disposal Containers
High Flow Solution System
Unit Size:
Each
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thread Size (Metric) Includes For Use With (Equipment) Unit Size
17298860 S60/61 Splash Guard, Sieve, Grounding Cable Disposal Containers Each
17142235 B83 Splash Guard, Sieve, Grounding Cable, Ball Valve, Lance Disposal Containers Each
17192225 GL45 Splash Guard, Sieve, Grounding Cable, Ball Valve, Lance Disposal Containers Each
17465053 GL45 Splash Guard, Sieve, Grounding Cable, Ball Valve, Lance High Flow Solution System Each
17288860 GL45 Splash Guard, 2 x Removable Sieves, Grounding Cable Disposal Containers Each
17102245 GL45 Splash Guard, Sieve, Grounding Cable, Ball Valve Disposal Containers Each
17172235 R2 Splash Guard, Sieve, Grounding Cable, Ball Valve Disposal Containers Each
17162235 S50 Splash Guard, Sieve, Grounding Cable, Ball Valve, Lance Disposal Containers Each
17112235 S51 Splash Guard, Sieve, Grounding Cable, Ball Valve, Lance Disposal Containers Each
17122235 S55 Splash Guard, Sieve, Grounding Cable, Ball Valve, Lance Disposal Containers Each
17182225 S60/61 Splash Guard, Sieve, Grounding Cable, Ball Valve, Lance Disposal Containers Each
17132235 S65 Splash Guard, Sieve, Grounding Cable, Ball Valve, Lance Disposal Containers Each
17152235 S70/71 Splash Guard, Sieve, Grounding Cable, Ball Valve, Lance Disposal Containers Each
17102235 S90 Splash Guard, Sieve, Grounding Cable, Ball Valve, Lance Disposal Containers Each
17192235 S95 Splash Guard, Sieve, Grounding Cable, Ball Valve, Lance Disposal Containers Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Product Code. 17298860 Supplier SCAT™ Supplier No. 317634
  • ONLINE EXCLUSIVE PRICE
    204.00€
    186.76€ / Pack of 1
    Save 17.24€
    8% Off
View all promotions
Estimated Shipment: 01-10-2026
Add to basket
Add to basket
The strong and electrically conductive safety funnel ARNOLD V2.0 with lid and ball meets the highest requirements for safe disposal of hazardous liquid waste due to its combination of two independent safety mechanisms.
The safety funnel ARNOLD V2.0 with lid and ball valve is available for different container threads. In order to be able to produce in an environmentally friendly and economical way, we supply individual variants with a thread adapter, which has no influence on the functionality or safety of the funnel.
  • Two safety features consist of a ball integrated in the funnel basin and a sealing ring incorporated in the hinged lid of the funnel, which prevent the escape of toxic or explosive gases
  • Electrically conductive HDPE material with grounding connection to prevent flying sparks
  • Hinged lid with sealing ring for closure when the funnel is not in use
  • Optimized sieve for collecting larger particles and for depositing laboratory bottles
  • Improved splash guard with integrated air channels to prevent bubble formation
  • Ball valve with automatic opening mechanism during filling
  • Individually shortenable safety lance for clean drainage of liquid waste
  • All major components are removable for easy cleaning
Specifications
Diameter (Metric)Top 200 mm (Funnel)
Capacity (Metric) 1500 mL
Material HDPE-EC
Height (Metric) 210 cm
Autoclavable Not Autoclavable
Color Black
For Use With (Application) Safe Disposal of Liquid and Flammable Wastes
For Use With (Equipment) Disposal Containers
Includes Splash Guard, Sieve, Grounding Cable
Thread Size (Metric) S60/61
Product Type Safety Funnel with Hinged Lid