missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ FTL (Human) Recombinant Protein
Human FTL full-length ORF ( AAH04245, 1 a.a. - 175 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00002512-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MSSQIRQNYSTDVEAAVNSLVNLYLQASYTYLSLGFYFDRDDVALEGVSHFFRELAEEKREGYERLLKMQNQRGGRALFQDIKKPAEDEWGKTPDAMKAAMALEKKLNQALLDLHALGSARTDPHLCDFLETHFLDEEVKLIKKMGDHLTNLHRLGGPEAGLGEYLFERLTLKHD
Specifications
AAH04245 | |
ferritin, light polypeptide | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FTL | |
GST |
2512 | |
Wheat germ expression system | |
25 μg | |
MGC71996 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |