Learn More
JULABO™ Immersion Cooler, FT Series
Product Code. p-8002185
Click to view available options
Model:
FT200
FT400
FT402
FT900
FT902
Temperature Range:
-20°C to +30°C
-40°C to +30°C
-90°C to +30°C
Unit Size:
Each
Product Code. 10334191
Brand: JULABO™ 9650820
Only null left
Add to basket
Edge
Add to basket
Primarily used for countercooling open bath circulators to ambient temperature or below as well as for rapidly cooling liquids. JULABO™ immersion coolers have a small footprint and represent an economic solution preventing tap water usage for cooling.
  • Rapid cooling of liquids down to low temperatures
  • Counter-cooling of liquids in combination with heating circulators
  • Environmentally friendly by saving precious tap water
  • User-friendly operation and handling
  • Compact design, small footprint
  • Dry ice substitution
Specifications
Ambient Temperature 5 to 40°C
Description Immersion Cooler FT
Cooling Capacity 0.25 kW at 20°C
Temperature Range -20°C to +30°C
Includes Immersion probe (L x dia.) 9 x 4 cm
Model FT200
Dimensions (L x W x H) 55 x 38 x 60 cm
Length (Metric) Hose 120 cm
Weight (Metric) 18 kg
Voltage 115/230 V