Learn More
Novus Biologicals™ FSH Native Protein
Product Code. 18027127 Shop All Bio Techne Products
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Product Code. 18027127
Brand: Novus Biologicals™ NBP19952550UG
Highly purified and high bioactivity. Generating reliable and reproducible results.
  • Native bioactive protein containing 165 amino acids for Human FSH
  • The FSH Native Protein is derived from Urine of post-menopausal women.
  • This product is for research use only and is not approved for use in humans or in clinical diagnosis.
TRUSTED_SUSTAINABILITY
Specifications
Concentration Lyoph
For Use With (Application) Bioactivity, SDS-PAGE
Molecular Weight (g/mol) 35.5 kDa
Purification Method >98%, by SDS-PAGE
Quantity 50 μg
Source Urine of post-menopausal women
Storage Requirements Store at -20 to -80°C. Avoid freeze-thaw cycles.
Endotoxin Concentration <0.1 ng/μg
Reconstitution Reconstitute in dH2O to a concentration of 0.1 - 1.0 mg/mL. The solution can then be diluted into other aqueous buffers and stored at 4°C for 1 week or -20°C for long-term storage.
Buffer Lyophilized.
Purity or Quality Grade >98%, by SDS-PAGE