Learn More
Thermo Scientific Chemicals FSB Buffer
Product Code. 15433059
Click to view available options
Quantity:
125 mL
250 mL
Unit Size:
125mL
250mL
Product Code. 15433059
Brand: Thermo Scientific Chemicals J60262.AK
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
For frozen storage of competent cells
For frozen storage of competent cells
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Buffer
Color Colorless
For Use With (Application) For frozen storage of competent cells
Physical Form Liquid
Quantity 250 mL
Synonym Frozen storage buffer