Learn More
Thermo Scientific™ Frosted Microscope Slides, Cut
Product Code. p-7112207 Shop All Epredia Products
Change view
Click to view available options
Dimensions (L x W):
75 x 50 mm
76 x 26 mm
Frosted:
20 mm
Fully Frosted
Quantity:
100 Pieces in Paper
50/Box, 1000 in outer carton
50/Box, 2000 in outer carton OEM
Writing Surface:
No
Yes
Unit Size:
Pack of 1000
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Frosted Quantity Writing Surface unitSize
12392138 76 x 26 mm 20 mm 50/Box, 2000 in outer carton OEM Yes Pack of 50
10782887 76 x 26 mm Fully Frosted 50/Box, 1000 in outer carton No Pack of 50
15363112 75 x 50 mm 20 mm 100 Pieces in Paper Yes Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12392138 Supplier Thermo Scientific™ Supplier No. AAAA000001##13E01FST20PH#
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Premium slides are manufactured through a unique proprietary cleaning process intended to provide consistent results and ease interpretation. This process allows these slides to to be used in routine histology and microbiology applications; and specifically in applications by delivering consistently crisp, featherededge blood smears with good morphology.
  • Manufactured from Swiss extra-white soda-lime glass
  • Reduced iron content for low self-fluorescence
  • Greenish shimmer minimized
  • Guaranteed optimum, faultless transparency
  • Clean, flat and suitable for immediate use
TRUSTED_SUSTAINABILITY
Specifications
Length (Metric) 76 mm
Width (Metric) 26 mm
Frosted 20 mm
Material Soda Lime Glass
Thickness (Metric) 1 mm
Certifications/Compliance ISO8037/I
Coating Uncoated
Quantity 50/Box, 2000 in outer carton OEM
Writing Surface Yes
Dimensions (L x W) 76 x 26 mm