Learn More
OHAUS™ Frontier™ 5513 Series Micro-Centrifuges
Product Code. 16221742 Shop All Ohaus Products
Change view
Click to view available options
Refrigerated:
No
Yes
Rotor Included:
No
Yes
Capacity:
18 x 1.5/2 mL Tubes
24 x 1.5/2 mL Tubes
Unit Size:
1 unit
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Refrigerated Rotor Included Capacity Unit Size
16221742 No Yes 24 x 1.5/2 mL Tubes 1 unit
16231742 No No 24 x 1.5/2 mL Tubes 1 unit
30130264 No Yes 18 x 1.5/2 mL Tubes Each
30130218 Yes No 24 x 1.5/2 mL Tubes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16221742 Supplier OHAUS™ Supplier No. 30370691
  • ONLINE EXCLUSIVE PRICE
    1845.00€
    1380.00€ / Pack of 1
    Save 465.00€
    25% Off
View all promotions
 Restricted Item
In Stock
Micro centrifuges have five optional rotors designed for clinical, industrial, and research applications. The FC5513 series features a uniquely designed splash-proof front panel with touch wheel control on an intuitive interface. Compact design saves valuable bench space in the lab.
Sleekly designed interface enables smooth operation for a great user experience
  • Easy-to-use touch-wheel interface for parameter adjustments
  • Convenient interface shows both set and actual parameters
  • One-touch toggling between RPM/RCF, Acc/Dec
  • Crisp display on a double-line LCD display with light blue backlight
Solid engineering to ensure functionality and reliability
  • True splash-proof panel protects the centrifuge from accidental sample spilling
  • Maintenance-free induction motor produces maximum G-force of 17317 x g for FC5513 and 17157 x g for FC5513R
  • Speeds up to 13500 rpm in less than 15 seconds with 24 x 1.5/2.0 mL rotor
  • 10 acceleration/deceleration settings
  • 99 memory sets to quickly store and recall commonly used parameters
  • FC5513R model has refrigeration capabilities from 40°C to -20°C
  • R290, an environmentally friendly coolant, contributes to the eco-friendliness of refrigerated centrifuges by minimizing their environmental impact and supporting efforts to combat global warming
  • Pre-cool feature in quickly lowers the temperature of rotor and chamber before a run
Flexible rotor selections to cover wide range of laboratory applications
  • Works with microtube rotors with different rotor capacity, 18 x 1.5/2.0 mL or 24 x 1.5/2.0 mL
  • Compatible with up to 7 rotor option and maximum rotor capacity of 24 x 1.5/2.0 mL
  • Support spin-column and hematocrit capillary tubes through additional rotor options
Safety features that prevent injury
  • Sealed and TÜV certified rotors to safeguard operators from harmful aerosols
  • Rotor imbalance sensing systems stops the unit when it detects an imbalanced load
  • Lid-locking system prevents centrifuge from opening during operation (FC5513R)
TRUSTED_SUSTAINABILITY
Specifications
Capacity 24 x 1.5/2 mL Tubes
Max. Speed 13,500 rpm
Max. RCF 17,317 x g
Height (English) Exterior 9.3 in.
Height (Metric) Exterior 236 mm
Length (English) Exterior 14 in.
Length (Metric) Exterior 356 mm
Noise Level ≤60±2 dBA
Width (English) Exterior 10.9 in.
Width (Metric) Exterior 277 mm
Certifications/Compliance CE, FCC
Safety Features CE, FCC
Electrical Requirements AC Power (Included)
Dimensions (L x W x H) Exterior 356 x 277 x 236 mm
Depth (English) 14 in.
Depth (Metric) 356 mm
For Use With (Equipment) 30210898, 30472302, 30472304, 83041014
Model FC5513+R02
Wattage 170 W
Maximum Force Range 17317 g
Rotor Included Yes
Refrigerated No
Run Time 10 sec. to 99 hr. 59 min. 59 sec., Continous, Quickspin
Dimensions (HWD) 236 x 277 x 356 mm
Features High sample throughput,multi-safety control, personal program function
Net Weight 13,1 kg
Voltage 230 V
Product Type Microcentrifuge
Display Type LCD with Backlight