Learn More
Carl Zeiss™ Front Lens System
Product Code. 11733724
Click to view available options
Magnification Power:
0.3X
Unit Size:
Each
Product Code. 11733724
Brand: Carl Zeiss™ 4550250000000
Only null left
Add to basket
Add to basket
For use with Stemi Microscopes
Compatible Microscopes:
  • Stemi 2000
  • Stemi 2000 C
  • Stemi 2000 CS
  • Stemi 508
  • Stemi 508 Demo
  • Stemi 508 doc
  • Stemi 508 trino
  • Stemi DV4 SPOT
Specifications
Type Front Lens System, FWD: 287 mm
Magnification Power 0.3X