Learn More
Wesbart™ Front Access Freezer Racks
Product Code. 15745855
Change view
Click to view available options
Dimensions (L x W x H):
560 x 140 x 415 mm
560 x 140 x 420 mm
560 x 145 x 235 mm
560 x 280 x 290 mm
590 x 165 x 420 mm
675 x 140 x 305 mm
Positions:
2
3
4
7
8
10
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) Positions unitSize
15745855 675 x 140 x 305 mm 3 Each
15729738 560 x 280 x 290 mm 2 Each
17152986 560 x 140 x 415 mm 3 Each
15737978 560 x 145 x 235 mm 4 Each
17162986 560 x 140 x 415 mm 4 Each
15739738 590 x 165 x 420 mm 7 Each
15751237 560 x 140 x 420 mm 8 Each
15719738 560 x 140 x 415 mm 10 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 15745855 Supplier Wesbart™ Supplier No. BH229/4A
Wesbart Front Access Freezer Racks are used to store boxes.
  • ULT freezer racks
  • For upright freezers to store boxes
Specifications
Autoclavable Autoclavable
Color Silver
Positions 3
Shape Rectangular
Stackable Yes
Sterility Non-sterile
For Use With (Application) Store Loose Items
For Use With (Equipment) Freezers
Dimensions (L x W x H) 675 x 140 x 305 mm
Product Type Freezer Rack