Learn More
TFA Dostmann™ Fridge Freezer Thermometer
Product Code. 11812602
Click to view available options
Temperature Range (Metric):
-40°C to +45°C
Unit Size:
Each
Product Code. 11812602
Brand: TFA Dostmann™ 14.4003.02.01
Only null left
Specifications
Product Type Fridge Thermometer
Max. Temperature (Metric) 45°C
Min. Temperature (Metric) -40°C
Temperature Range (Metric) -40°C to +45°C