Learn More
Labconco™ FreeZone Benchtop Shell Freezer, 230 V, Schuko Plug and Receptacle
Shop All Labconco Corporation Products
Click to view available options
Capacity (Metric):
12 L
Unit Size:
Each
Description
- 1/2 hp refrigeration system prefreezes samples to -40 °C
- Stainless-steel bath with two rollers to rotate flasks in low-temperature heat transfer solution
- Produces uniform thin coating of sample on container interior wall
- Ensures efficient vapor flow from sample to collector during lyophilization
- Produces uniform thin coating of sample on container interior wall
- Accepts Fast-Freeze flasks up to 1200 mL in size
- Does not contain ozone-depleting chlorofluorocarbons (CFCs)
- LED display of bath temperature from -20 to -42 °C in predetermined increments
- Left-mounted retractable bath hose facilitates cleaning of bath
- Fits on countertop or lab cart
- CE conformity (230 V, 50 Hz model)
- White powder-coated steel cabinet with brushed stainless-steel front and blue accents
- Stainless-steel bath with urethane foam insulation and white HDPE cover
Specifications:
- Shipping weight: 44 kg (96 lb.)
- Cabinet dimensions (L x W x H): 56.1 x 38.4 x 50 cm (22.1 x 15.1 x 19.7 in.)
- Bath dimensions (D x W x H): 30.5 x 14 x 16.5 cm (12 x 5.5 x 6.5 in.)
Specifications
Specifications
| Type | Fast-Freeze Flask Shell Freezer |
| Capacity (Metric) | 12 L |
| Capacity (English) | 0.42 feet3 |
| Temperature Range (Metric) | -40°C |
| Temperature Range (English) | -40°F |
| Material | Stainless Steel |
| Certifications/Compliance | CE |
| For Use With (Equipment) | Freeze dryers |
| Dimensions (L x W x H) Exterior | 56.1 x 36.8 x 50 cm |
| Depth (English) | 22.1 in. |
| Show More |
Product Content Correction
Product Title
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto